missing translation for 'onlineSavingsMsg'
Learn More

Methyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog Number p-7050385
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
553-97-9
C7H6O2
122.12
MFCD00001603
VTWDKFNVVLAELH-UHFFFAOYSA-N
p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinone
11122
2-methylcyclohexa-2,5-diene-1,4-dione
CC1=CC(=O)C=CC1=O
This item is not returnable. View return policy
553-97-9
C7H6O2
122.12
MFCD00001603
VTWDKFNVVLAELH-UHFFFAOYSA-N
p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinone
11122
2-methylcyclohexa-2,5-diene-1,4-dione
CC1=CC(=O)C=CC1=O

Methyl-p-benzoquinone imine is the products were the tetrahedral adducts. Synthesis of 2H-indazole-4,7-dione derivatives from3-phenylsydnone and p-toluquinone. A Michael addition of 2-methylcyclopentane-1,3-dione to various p-toluquinone imides exclusively afforded the sole products in a regiospecific manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 67°C to 70°C
Color Green-Yellow
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 122.12
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Methyl-p-benzoquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'