missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Orange, ACS
19.67 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Beschreibung
Methyl orange is a pH indicator frequently used in titrations, also used for histological microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Melting Point | >300°C |
UN Number | UN3143 |
Beilstein | 4732884 |
Merck Index | 14,6105 |
Solubility Information | Soluble in 500 parts water; More soluble in hot water |
Formula Weight | 327.34 |
Grade | ACS |
Odor | Weak, characteristic |
Color | Orange |
pH | 6.5 |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'