missing translation for 'onlineSavingsMsg'
Learn More

Methyl nonadecafluorodecanoate, 94%, ACROS Organics™

Brand:  Acros Organics 342230250

Code : 29

Additional Details : CAS Number : 307-79-9 Weight : 0.09400kg


Quantity 25mL
Packaging Glass Bottle

 View more versions of this product

Product Code. 10245181

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Methyl nonadecafluorodecanoate
307-79-9
94%
CF3(CF)8COCH3
MFCD00153211
94%
≥92%
C11H3F19O2
528.11

Specifications

Methyl nonadecafluorodecanoate
94%
Authentic
Glass Bottle
≥92%
C11H3F19O2
528.11
1.76g/mL
25mL
307-79-9
1.3070 to 1.3090 (20°C, 589nm)
Liquid
94%
CF3(CF)8COCH3
MFCD00153211
Clear colorless
Videos
SDS