missing translation for 'onlineSavingsMsg'
Learn More

Methyl nicotinate, 99%, Thermo Scientific Chemicals

Catalog Number p-165480
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
93-60-7
C7H7NO2
137.14
MFCD00006388
YNBADRVTZLEFNH-UHFFFAOYSA-N
methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
7151
methyl pyridine-3-carboxylate
COC(=O)C1=CN=CC=C1
This item is not returnable. View return policy
93-60-7
C7H7NO2
137.14
MFCD00006388
YNBADRVTZLEFNH-UHFFFAOYSA-N
methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
7151
methyl pyridine-3-carboxylate
COC(=O)C1=CN=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38.0°C to 43.0°C
Color Brown to White
Boiling Point 209.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 22, 39
Merck Index 15, 6173
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol and benzene
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl nicotinate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl nicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'