missing translation for 'onlineSavingsMsg'
Learn More

Methyl isovalerate, 98%

Product Code. 11322436
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11322436

Brand: Thermo Scientific Alfa Aesar L06241.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-24-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-24-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00042866
InChI Key OQAGVSWESNCJJT-UHFFFAOYSA-N
Synonym methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
PubChem CID 11160
IUPAC Name methyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC

Specifications

Density 0.88
Boiling Point 115°C to 117°C
Flash Point 16°C (60°F)
Packaging Glass bottle
Refractive Index 1.3936
Quantity 50 g
UN Number UN2400
Beilstein 1699922
Specific Gravity 0.88
Formula Weight 116.17
Percent Purity 98%
Chemical Name or Material Methyl isovalerate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.