missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl isothiocyanate, 98%

Brand:  Alfa Aesar™ A11757.14

Code : NEW

Additional Details : CAS Number : 556-61-6 Weight : 0.01000kg

Quantity 25g

 View more versions of this product

Product Code. 11491880

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Methyl isothiocyanate
CH3NCS
MFCD00004818
605319
556-61-6
73.12
UN2477
14,6092

Specifications

Methyl isothiocyanate
Fused Solid
CH3NCS
MFCD00004818
605319
117° to 118°C
25g
556-61-6
98%
73.12
UN2477
14,6092
31° to 36°C
30°C (86°F)
Videos
SDS