missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-5-carboxylate, 99%, Thermo Scientific™

Catalog Number p-3770804
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1011-65-0
C10H9NO2
175.19
MFCD00153023
DRYBMFJLYYEOBZ-UHFFFAOYSA-N
methyl indole-5-carboxylate, indole-5-carboxylic acid methyl ester, 1h-indole-5-carboxylic acid, methyl ester, methylindole-5-carboxylate, 1h-indole-5-carboxylic acid methyl ester, indole5carboxylicacidmethylester, indole-5-carboxylicacidmethylester, indole-5-carboxylate, pubchem7248, 5-methoxycarbonylindole
2737635
methyl 1H-indole-5-carboxylate
COC(=O)C1=CC2=C(C=C1)NC=C2
This item is not returnable. View return policy
1011-65-0
C10H9NO2
175.19
MFCD00153023
DRYBMFJLYYEOBZ-UHFFFAOYSA-N
methyl indole-5-carboxylate, indole-5-carboxylic acid methyl ester, 1h-indole-5-carboxylic acid, methyl ester, methylindole-5-carboxylate, 1h-indole-5-carboxylic acid methyl ester, indole5carboxylicacidmethylester, indole-5-carboxylicacidmethylester, indole-5-carboxylate, pubchem7248, 5-methoxycarbonylindole
2737635
methyl 1H-indole-5-carboxylate
COC(=O)C1=CC2=C(C=C1)NC=C2

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 126°C to 128°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material Methyl indole-5-carboxylate
compliance-icons
Product Identifier
  • Methyl indole-5-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'