Learn More
H3K56me1 Recombinant Rabbit Monoclonal Antibody (RM180), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA524682
Additional Details : Weight : 0.01000kg
Description
This antibody reacts to Histone H3 monomethylated at Lysine 56 (K56me1). No cross reactivity with nonmodified Lysine 56 or other methylations in histone H3. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
H3K56me1 Recombinant Monoclonal antibody specifically detects H3K56me1 in Human samples. It is validated for ELISA, Luminex, Western Blot.Specifications
H3K56me1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4 | |
P68431, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7 | |
Monomethyl-peptide corresponding to Monomethyl-Histone H3 (Lys56). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Luminex, Western Blot | |
RM180 | |
Unconjugated | |
H3C1 | |
B0035.7; CELE_T10C6.12; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; h3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K56; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-53; H3-614; H3a; H3b; H3-B; H3c1; H3c10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3f; H3-F; H3F1K; H3F2; H3F3; H3F3A; H3f3b; H3F3C; H3FA; H3F | |
Rabbit | |
Protein A | |
RUO | |
126961, 333932, 8350, 8356, 8357, 8358, 8968 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
For Research Use Only