Learn More
Methyl glycolate, 98%, Thermo Scientific Chemicals
Description
Methyl glycolate is a chemical reagent used in the synthesis of taxol and rapamycin. It is used in leather tanning and textile dyeing. It acts as a flavoring agent, preservative in food processing, in water treatment industries and as chemical intermediates. Further, it is used in emulsion polymers, ink and paint additive to improve flow properties and impart gloss. It plays an important role in producing triisopropylsilanyloxy-acetic acid methyl ester using imidazole as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl glycolate is a chemical reagent used in the synthesis of taxol and rapamycin. It is used in leather tanning and textile dyeing. It acts as a flavoring agent, preservative in food processing, in water treatment industries and as chemical intermediates. Further, it is used in emulsion polymers, ink and paint additive to improve flow properties and impart gloss. It plays an important role in producing triisopropylsilanyloxy-acetic acid methyl ester using imidazole as reagent.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Density | 1.184 |
Boiling Point | 150°C to 151°C |
Flash Point | 67°C (152°F) |
Assay Percent Range | 98+% |
Linear Formula | HOCH2CO2CH3 |
Refractive Index | 1.417 |
Quantity | 5 g |
Beilstein | 1699571 |
Solubility Information | Miscible with water. |
Formula Weight | 90.08 |
Show More |
RUO – Research Use Only