missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 97%, pure, ACROS Organics™

Brand:  Acros Organics 126820100

Code : NEW

Additional Details : CAS Number : 107-31-3 Weight : 10.00000kg Transport : UN number : 1243 Chem class : 3 Pack group : I

Quantity 10L
Packaging Metal drum

 View more versions of this product

Product Code. 13408348

  • £108.70 / 10 L


Estimated Shipment
18-06-2018
Add to basket

Specifications

Chemical Identifiers

Methyl formate
107-31-3
97%
C2H4O2
60.05
Formic acid methyl ester, Methyl methanoate
97%
Pure
97%
HCO2CH3
MFCD00003291

Specifications

Methyl formate
0.968
Clear colorless
10L
107-31-3
Authentic
1.3420 to 1.3440
Liquid
97%
0.05% max.
60.05
Formic acid methyl ester, Methyl methanoate
−32°C
31.0° to 33.0°C
− 100.0°C
97%
0.9680g/mL
Metal drum
Pure
97%
C2H4O2
HCO2CH3
MFCD00003291
Videos