missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Methyl-DL-phenylalanine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 284°C to 286°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 5 g |
Formula Weight | 179.22 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | α-Methyl-DL-phenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'