Learn More
Methyl chloroacetate, 99+%, Thermo Scientific Chemicals
Description
Methyl chloroacetate is used as a solvent and chemical intermediate. It acts as a precursor in the preparation of (carboxymethyl) trimethylammonium chloride esters. Further, it is used in the preparation of octakis-(carbethoxymethoxy)calix[8]arene. It is employed as an extraction solvent during the separation of neutral compounds. In addition to this, it is used in the synthesis of dimethyl carbonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl chloroacetate is used as a solvent and chemical intermediate. It acts as a precursor in the preparation of (carboxymethyl) trimethylammonium chloride esters. Further, it is used in the preparation of octakis-(carbethoxymethoxy)calix[8]arene. It is employed as an extraction solvent during the separation of neutral compounds. In addition to this, it is used in the synthesis of dimethyl carbonate.
Solubility
Miscible with organic solvents, alcohol, ether, acetone and benzene. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | -32°C |
Density | 1.236 |
Boiling Point | 128°C to 130°C |
Flash Point | 51°C (123°F) |
Refractive Index | 1.422 |
Quantity | 250 g |
UN Number | UN2295 |
Beilstein | 506255 |
Merck Index | 14,6042 |
Solubility Information | Miscible with organic solvents,alcohol,ether,acetone and benzene. Slightly miscible with water. |
Show More |
RUO – Research Use Only