missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cPs

Catalog Number p-7050404
Click to view available options
Quantity:
100 g
500 g
2.5 kg
9004-67-5
C20H38O11
454.513
MFCD00081763
YLGXILFCIXHCMC-JHGZEJCSSA-N
51063134
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
9004-67-5
C20H38O11
454.513
MFCD00081763
YLGXILFCIXHCMC-JHGZEJCSSA-N
51063134
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Methyl cellulose is used as a thickener and an emulsifier in various food and cosmetic products; as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. In paper and textile industries, it is used as a sizing, thereby protecting the fibers from water or oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 g
Merck Index 14,6040
Solubility Information Soluble in cold water.
Physical Form Powder
Chemical Name or Material Methyl cellulose, Viscosity: 4000cPs

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'