missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-bromophenylacetate, 97+%, ACROS Organics™

Brand:  Acros Organics 379300010

Code : 29

Additional Details : CAS Number : 3042-81-7 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 1a Pack group : II


Quantity 1g

 View more versions of this product

Product Code. 10623975

  • £10.50 / 1g


Estimated Shipment
19-03-2018
Add to basket


Specifications

Chemical Identifiers

Methyl α-bromophenylacetate
C9H9BrO2
229.07
≥97%
C6H5CH(Br)CO2CH3
MFCD00013535

Specifications

Methyl α-bromophenylacetate
1.4550g/mL
97+%
C9H9BrO2
1.455
MFCD00013535
113°C
3042-81-7
Glass bottle
≥97%
C6H5CH(Br)CO2CH3
229.07
113.0°C (3.0 mmHg)
1g
Videos
SDS