missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-bromophenylacetate, 97+%, ACROS Organics™

Brand:  Acros Organics 379300050

Code : 29

Additional Details : CAS Number : 3042-81-7 Weight : 0.05500kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Quantity 5g

 View more versions of this product

Product Code. 10537244

  • £29.00 / 5g


Estimated Shipment
20-07-2018
Add to basket


Specifications

Chemical Identifiers

Methyl α-bromophenylacetate
C9H9BrO2
229.07
≥97%
C6H5CH(Br)CO2CH3
MFCD00013535

Specifications

Methyl α-bromophenylacetate
1.4550g/mL
97+%
C9H9BrO2
1.455
MFCD00013535
113°C
3042-81-7
Glass bottle
≥97%
C6H5CH(Br)CO2CH3
229.07
113.0°C (3.0 mmHg)
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl alpha-bromophenylacetate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS