missing translation for 'onlineSavingsMsg'
Learn More

(Methyl benzoate)tricarbonylchromium, 98%, Thermo Scientific™

Catalog Number p-7059307
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
12125-87-0
C11H8CrO5
272.18
MFCD00009957
GEALMZJLYGNHRA-UHFFFAOYSA-N
methyl benzoate tricarbonylchromium, tricarbonyl methyl benzoate chromium
11780645
[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].COC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
12125-87-0
C11H8CrO5
272.18
MFCD00009957
GEALMZJLYGNHRA-UHFFFAOYSA-N
methyl benzoate tricarbonylchromium, tricarbonyl methyl benzoate chromium
11780645
[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].COC(=O)C1=CC=CC=C1

Catalyst for hydrogenation of unsaturated fats

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 95°C to 96°C
Color Orange
Quantity 1 g
UN Number UN3466
Sensitivity Air and moisture sensitive
IUPAC Name tris(methanidylidyneoxidanium) methyl benzoate chromium
Formula Weight 272.18
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material (Methyl benzoate)tricarbonylchromium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'