missing translation for 'onlineSavingsMsg'
Learn More

Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™

Catalog Number p-3486250
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4338-95-8
C2H7IN2S
218.06
MFCD00035598
LFXAECSQJSRSTP-UHFFFAOYSA-N
methyl aminomethanimidothioate hydroiodide, sulfiode, methylthiouronium iodide, methyl carbamimidothioate hydroiodide, s-methylthiuronium iodide, s-methylthiouronium iodide, s-methylisothiouronium iodide, 2-methyl-2-thiopseudourea hydroiodide, 2-methylisothiouronium iodide, s-methylisothiourea hydriodide
197812
[H+].[I-].CSC(N)=N
This item is not returnable. View return policy
4338-95-8
C2H7IN2S
218.06
MFCD00035598
LFXAECSQJSRSTP-UHFFFAOYSA-N
methyl aminomethanimidothioate hydroiodide, sulfiode, methylthiouronium iodide, methyl carbamimidothioate hydroiodide, s-methylthiuronium iodide, s-methylthiouronium iodide, s-methylisothiouronium iodide, 2-methyl-2-thiopseudourea hydroiodide, 2-methylisothiouronium iodide, s-methylisothiourea hydriodide
197812
[H+].[I-].CSC(N)=N
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl carbamimidothioate;hydroiodide
Formula Weight 218.06
Percent Purity 95%
Chemical Name or Material Methyl aminomethanimidothioate hydroiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'