missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl alpha-bromo-4-chlorophenylacetate, 97%

Brand:  Alfa Aesar™ H64969.03

Code : NEW

Additional Details : CAS Number : 24091-92-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15492399

  • £62.75 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl alpha-bromo-4-chlorophenylacetate
263.51
1529651
24091-92-7
MFCD16038767
α-Bromo-4-chlorophenylacetic acid methyl ester

Specifications

Methyl alpha-bromo-4-chlorophenylacetate
Liquid
263.51
Colorless
1g
24091-92-7
97%
MFCD16038767
1529651
α-Bromo-4-chlorophenylacetic acid methyl ester
Videos
SDS