missing translation for 'onlineSavingsMsg'
Learn More

Methyl acetate, 99+%, Extra Dry, AcroSeal™, ACROS Organics™

£21.50 - £185.00

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10037772
SDS View Documents
Acros Organics
371831000
100mL
£21.50
100mL
View alternative products
Save Up to 
»
In stock, ready to ship: 31
Please to purchase this item. Need a web account? Register with us today!
10195512
SDS View Documents
Acros Organics
371830010
1L
£86.70
1 L
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
10759873
SDS View Documents
Acros Organics
371830025
2.5L
£185.00
2.50 L
View alternative products
Save Up to 
»
In stock, ready to ship: 2
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

79-20-9
74.08
KXKVLQRXCPHEJC-UHFFFAOYSA-N
6584
methyl acetate
C3H6O2
MFCD00008711
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
CHEBI:77700
CC(=O)OC

Specifications

Methyl acetate
−10°C
AcroSeal glass Bottle
0.0005 % max.
0.38 mPa.s (20°C)
57.4°C
-98.0°C
79-20-9
C3H6O2
MFCD00008711
15, 6082
Solubility in water: 250g/L (20°C). Other solubilities: miscible with alcohol and ether
CC(=O)OC
74.08
CHEBI:77700
Solution
Extra Dry
0.9300g/mL
Authentic
1.3600 to 1.3620
0.93
0.005% Max.
Colorless to Yellow
100mL
99% min. (GC)
CH3CO2CH3
02, 224
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
KXKVLQRXCPHEJC-UHFFFAOYSA-N
methyl acetate
6584
74.08
99+%
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl acetate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications