missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 6-(hydroxymethyl)pyridine-2-carboxylate, 95%

Brand:  Alfa Aesar™ H63793.06

Code : NEW

Additional Details : CAS Number : 39977-44-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438526

  • £399.90 / 5g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 6-(hydroxymethyl)pyridine-2-carboxylate
167.16
6-(Hydroxymethyl)pyridine-2-carboxylic acid methyl ester
39977-44-1
MFCD03840850

Specifications

Methyl 6-(hydroxymethyl)pyridine-2-carboxylate
5g
Solid
167.16
6-(Hydroxymethyl)pyridine-2-carboxylic acid methyl ester
White
39977-44-1
95%
MFCD03840850
Videos
SDS