missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 6-(hydroxymethyl)nicotinate, 98%

Brand:  Alfa Aesar™ H64750.03

Code : NEW

Additional Details : CAS Number : 56026-36-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472069

  • £140.98 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 6-(hydroxymethyl)nicotinate
167.16
473391
56026-36-9
MFCD00643821
6-(Hydroxymethyl)nicotinic acid methyl ester; 6-Hydroxymethylpyridine-3-carboxylate methyl ester

Specifications

Methyl 6-(hydroxymethyl)nicotinate
75° to 78°C
56026-36-9
98%
MFCD00643821
6-(Hydroxymethyl)nicotinic acid methyl ester; 6-Hydroxymethylpyridine-3-carboxylate methyl ester
Off-white
1g
Solid
167.16
473391
Videos
SDS