missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-fluoronicotinate, 95%, Thermo Scientific Chemicals

Catalog Number 15477818
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1427-06-1
C7H6FNO2
155.128
MFCD03095098
HLYBWNNPVXFCPZ-UHFFFAOYSA-N
13758765
methyl 6-fluoropyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)F
This item is not returnable. View return policy
1427-06-1
C7H6FNO2
155.128
MFCD03095098
HLYBWNNPVXFCPZ-UHFFFAOYSA-N
13758765
methyl 6-fluoropyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 48°C to 52°C
Flash Point 99°C (210°F)
Quantity 250 mg
Synonym methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate
Formula Weight 155.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 6-fluoronicotinate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'