missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-oxopyrrolidine-2-carboxylate, 95%, Thermo Scientific™

Catalog Number 15442169
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
54571-66-3
C6H9NO3
143.14
MFCD03844651
HQGPKMSGXAUKHT-UHFFFAOYNA-N
methyl dl-pyroglutamate, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, methyl 5-oxo-2-pyrrolidinecarboxylate, 5-oxo-2-pyrrolidinecarboxylic acid methyl ester, methyl 5-oxo-dl-prolinate, methyl 5-oxo-2-pyrrolidinecarboxylate #, proline, 5-oxo-, methylester, 5-oxo-dl-proline methyl ester, methyl 2-pyrolidone-5-carboxylate, dl-pyroglutamic acid methyl ester
500249
methyl 5-oxopyrrolidine-2-carboxylate
COC(=O)C1CCC(=O)N1
This item is not returnable. View return policy
54571-66-3
C6H9NO3
143.14
MFCD03844651
HQGPKMSGXAUKHT-UHFFFAOYNA-N
methyl dl-pyroglutamate, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, methyl 5-oxo-2-pyrrolidinecarboxylate, 5-oxo-2-pyrrolidinecarboxylic acid methyl ester, methyl 5-oxo-dl-prolinate, methyl 5-oxo-2-pyrrolidinecarboxylate #, proline, 5-oxo-, methylester, 5-oxo-dl-proline methyl ester, methyl 2-pyrolidone-5-carboxylate, dl-pyroglutamic acid methyl ester
500249
methyl 5-oxopyrrolidine-2-carboxylate
COC(=O)C1CCC(=O)N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 21°C to 23°C (literature)
Boiling Point 133°C to 135°C (1 mmHg)
Refractive Index 1.485
Quantity 1 g
Beilstein 81950
Formula Weight 143.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 5-oxopyrrolidine-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'