missing translation for 'onlineSavingsMsg'
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech., Thermo Scientific Chemicals

Catalog Number p-7081135
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
25134-21-8
C10H10O3
178.19
MFCD00167036
KNRCVAANTQNTPT-UHFFFAOYNA-N
Methyl nadic anhydride
This item is not returnable. View return policy
25134-21-8
C10H10O3
178.19
MFCD00167036
KNRCVAANTQNTPT-UHFFFAOYNA-N
Methyl nadic anhydride

Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

Solubility
Miscible with acetone, benzene, naphtha and xylene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.232
Boiling Point 137°C to 140°C (10 mmHg)
Flash Point 135°C (275°F)
Refractive Index 1.5052
Quantity 100 g
Sensitivity Moisture sensitive
Solubility Information Miscible with acetone,benzene,naphtha and xylene.
Formula Weight 178.19
Grade Technical
Chemical Name or Material Methyl-5-norbornene-2,3-dicarboxylic anhydride, Mixture of isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'