missing translation for 'onlineSavingsMsg'
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, Thermo Scientific Chemicals

Catalog Number 10472791
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
25134-21-8
MFCD00167036
KNRCVAANTQNTPT-UHFFFAOYNA-N
Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride
This item is not returnable. View return policy
25134-21-8
MFCD00167036
KNRCVAANTQNTPT-UHFFFAOYNA-N
Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.2300g/mL
Boiling Point 275.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C10H10O3
Refractive Index 1.5060 to 1.5080
Quantity 250 g
Beilstein 17, 461
Specific Gravity 1.23
Solubility Information Solubility in water: <1g/L
Molecular Weight (g/mol) 178.18
Viscosity 220-300 mPa.s (25°C)
Formula Weight 178.18
Percent Purity 95%
Physical Form Oily Liquid
Chemical Name or Material Methyl-5-norbornene-2, 3-dicarboxylic anhydride, Mixture of isomers
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'