missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methylsalicylate 97%, ACROS Organics™

Brand:  Acros Organics 133990050

Code : 29

Additional Details : CAS Number : 22717-57-3 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10691633

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Methyl 5-methylsalicylate
22717-57-3
97%
CH3C6H3(OH)COOCH3
MFCD00016485
97%
97%
C9H10O3
166.18

Specifications

Methyl 5-methylsalicylate
22717-57-3
Glass bottle
Liquid
97%
CH3C6H3(OH)COOCH3
MFCD00016485
5g
97%
Authentic
1.5320 to 1.5340
97%
C9H10O3
166.18
Clear slightly yellow
Videos
SDS