missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-hexynoate, 95%, Thermo Scientific Chemicals

Catalog Number p-3586451
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
77758-51-1
C7H10O2
126.15
MFCD00671366
LZULAZTXJLWELL-UHFFFAOYSA-N
methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester
534077
methyl hex-5-ynoate
COC(=O)CCCC#C
This item is not returnable. View return policy
77758-51-1
C7H10O2
126.15
MFCD00671366
LZULAZTXJLWELL-UHFFFAOYSA-N
methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester
534077
methyl hex-5-ynoate
COC(=O)CCCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 60°C to 62°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CHC(CH2)3COOCH3
Refractive Index 1.4330 to 1.435
Quantity 5 g
Formula Weight 126.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 5-hexynoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'