missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-(chloromethyl)-2-furoate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 174.58 |
Percent Purity | 95% |
Chemical Name or Material | Methyl 5-(chloromethyl)-2-furoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'