missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-(chloromethyl)-2-furancarboxylate, 99%, Thermo Scientific™

Catalog Number p-2691757
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2144-37-8
174.58
MFCD00020922
PWXMEBZOKUPCST-UHFFFAOYSA-N
methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
75065
methyl 5-(chloromethyl)furan-2-carboxylate
COC(=O)C1=CC=C(O1)CCl
This item is not returnable. View return policy
2144-37-8
174.58
MFCD00020922
PWXMEBZOKUPCST-UHFFFAOYSA-N
methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
75065
methyl 5-(chloromethyl)furan-2-carboxylate
COC(=O)C1=CC=C(O1)CCl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 31°C to 35°C
Color White to Yellow
Boiling Point 102.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7ClO3
Quantity 1 g
Formula Weight 174.58
Percent Purity 99%
Physical Form Oily Liquid or Low Melting Crystalline Solid
Chemical Name or Material Methyl 5-(chloromethyl)-2-furancarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 5-(chloromethyl)-2-furancarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'