missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 5-bromoorotate, 97%

Brand:  Alfa Aesar™ H64761.06

Code : NEW

Additional Details : CAS Number : 30825-88-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15462099

  • £289.90 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 5-bromoorotate
249.02
915036
30825-88-8
MFCD07023034
5-Bromoorotic acid methyl ester; Methyl 5-bromo-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylate

Specifications

Methyl 5-bromoorotate
Solid
249.02
915036
252° to 254°C
30825-88-8
97%
MFCD07023034
5-Bromoorotic acid methyl ester; Methyl 5-bromo-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylate
5g
Videos
SDS