missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-6-chloronicotinate, 98%, Thermo Scientific Chemicals

Catalog Number p-7056017
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
78686-77-8
C7H5BrClNO2
250.476
MFCD03844855
WINGWVOUOFMOJQ-UHFFFAOYSA-N
2764313
methyl 5-bromo-6-chloropyridine-3-carboxylate
COC(=O)C1=CC(=C(N=C1)Cl)Br
This item is not returnable. View return policy
78686-77-8
C7H5BrClNO2
250.476
MFCD03844855
WINGWVOUOFMOJQ-UHFFFAOYSA-N
2764313
methyl 5-bromo-6-chloropyridine-3-carboxylate
COC(=O)C1=CC(=C(N=C1)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 79°C
Quantity 1 g
Beilstein 4403950
Synonym methyl 5-bromo-6-chloronicotinate, 5-bromo-6-chloro-nicotinic acid methyl ester, methyl-5-bromo-6-chloronicotinate, 3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester, methyl 3-bromo-2-chloropyridine-5-carboxylate, zlchem 1175, 3-bromo-4-chloro-benzoicacidmethylester, pubchem12980, acmc-1bdei, methyl5-bromo-6-chloronicotinate
Formula Weight 250.48
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 5-bromo-6-chloronicotinate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'