missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-bromo-2-(methylsulfanyl)-4-pyrimidinecarboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate |
Formula Weight | 263.11 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 5-bromo-2-(methylsulfanyl)-4-pyrimidinecarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'