missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 5-bromo-2-methylbenzoate, 98%

Brand:  Alfa Aesar™ H61861.14

Code : NEW

Additional Details : CAS Number : 79669-50-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15428448

  • £27.40 / 25g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Description

Description

Applications
Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

Methyl 5-bromo-2-methylbenzoate
229.07
5-Bromo-2-methylbenzoic acid methyl ester
79669-50-4
MFCD09839555

Specifications

Methyl 5-bromo-2-methylbenzoate
Powder
229.07
5-Bromo-2-methylbenzoic acid methyl ester
25g
79669-50-4
98%
MFCD09839555
Pale yellow
Videos
SDS