missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 5-bromo-2-chloronicotinate, 98%

Brand:  Alfa Aesar™ H64791.06

Code : NEW

Additional Details : CAS Number : 78686-79-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15452149

  • £14.50 / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

Methyl 5-bromo-2-chloronicotinate
250.48
4394169
78686-79-0
MFCD03844848
5-Bromo-2-chloronicotinic acid methyl ester; Methyl 5-bromo-2-chloropyridine-3-carboxylate

Specifications

Methyl 5-bromo-2-chloronicotinate
Solid
250.48
4394169
48° to 53°C
Light, Air & Moisture Sensitive
78686-79-0
98%
MFCD03844848
5-Bromo-2-chloronicotinic acid methyl ester; Methyl 5-bromo-2-chloropyridine-3-carboxylate
5g
Videos
SDS