missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 286.27 |
Percent Purity | 95% |
Chemical Name or Material | Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'