missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-oxotetrahydrothiophene-3-carboxylate, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Formula Weight | 160.19 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Methyl 4-oxotetrahydrothiophene-3-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'