missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methylsalicylate, 98+%, Thermo Scientific Chemicals

Catalog Number 15412159
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4670-56-8
C9H10O3
166.176
MFCD00020130
UITFCFWKYAOJEJ-UHFFFAOYSA-N
methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate
78400
methyl 2-hydroxy-4-methylbenzoate
CC1=CC(=C(C=C1)C(=O)OC)O
This item is not returnable. View return policy
4670-56-8
C9H10O3
166.176
MFCD00020130
UITFCFWKYAOJEJ-UHFFFAOYSA-N
methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate
78400
methyl 2-hydroxy-4-methylbenzoate
CC1=CC(=C(C=C1)C(=O)OC)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 27°C to 28°C
Color Brown to Yellow
Density 1.147 g/mL
Boiling Point 242°C (literature)
Refractive Index 1.537
Quantity 25 g
Beilstein 2690247
Formula Weight 166.18
Percent Purity ≥98%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 4-methylsalicylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'