missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-iodo-3-methylbenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.
Solubility
Slightly soluble in methanol.
Notes
Light sensitive. Store away from oxidizing agents, light.
Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.
Solubility
Slightly soluble in methanol.
Notes
Light sensitive. Store away from oxidizing agents, light.
Specifications
Melting Point | 59°C to 62°C |
Quantity | 5 g |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in methanol. |
Formula Weight | 276.07 |
Percent Purity | 98% |
Chemical Name or Material | Methyl 4-iodo-3-methylbenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'