missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 4-hydroxyindole-6-carboxylate, 97%

Brand:  Alfa Aesar™ H63398.03

Code : NEW

Additional Details : CAS Number : 77140-48-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400079

  • £233.20 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Description

Description

Applications
Methyl 4-hydroxyindole-6-carboxylate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (2.5 g/L) (25°C).

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

Methyl 4-hydroxyindole-6-carboxylate
191.18
4-Hydroxyindole-6-carboxylic acid methyl ester
77140-48-8
MFCD04973985

Specifications

Methyl 4-hydroxyindole-6-carboxylate
1g
Solid
191.18
4-Hydroxyindole-6-carboxylic acid methyl ester
Pale pink
77140-48-8
97%
MFCD04973985
Videos
SDS