missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4'-formylbiphenyl-4-carboxylate, 97%, Thermo Scientific™

Catalog Number 11995111
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
70916-89-1
C15H12O3
240.26
MFCD04039123
URINKTWOCVLGGR-UHFFFAOYSA-N
4'-formylbiphenyl-4-carboxylic acid methyl ester, methyl 4'-formyl-1,1'-biphenyl-4-carboxylate, methyl 4-4-formylphenyl benzoate, methyl 4'-formyl 1,1'-biphenyl-4-carboxylate, acmc-209ogh, methyl 4'-formylbiphenyl-4-carboxylate, methyl-4'-formylbiphenyl-4-carboxylate, 1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester, 1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester
4321323
methyl 4-(4-formylphenyl)benzoate
COC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
70916-89-1
C15H12O3
240.26
MFCD04039123
URINKTWOCVLGGR-UHFFFAOYSA-N
4'-formylbiphenyl-4-carboxylic acid methyl ester, methyl 4'-formyl-1,1'-biphenyl-4-carboxylate, methyl 4-4-formylphenyl benzoate, methyl 4'-formyl 1,1'-biphenyl-4-carboxylate, acmc-209ogh, methyl 4'-formylbiphenyl-4-carboxylate, methyl-4'-formylbiphenyl-4-carboxylate, 1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester, 1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester
4321323
methyl 4-(4-formylphenyl)benzoate
COC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1
Melting Point 113.0°C to 118.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 240.26
Percent Purity 97%
Chemical Name or Material Methyl 4'-formylbiphenyl-4-carboxylate
compliance-icons
Product Identifier
  • Methyl 4'-formylbiphenyl-4-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'