missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbenzoate, 99%, Thermo Scientific Chemicals

Catalog Number p-163329
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1571-08-0
C9H8O3
164.16
MFCD00006950
FEIOASZZURHTHB-UHFFFAOYSA-N
methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
15294
methyl 4-formylbenzoate
COC(=O)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
1571-08-0
C9H8O3
164.16
MFCD00006950
FEIOASZZURHTHB-UHFFFAOYSA-N
methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
15294
methyl 4-formylbenzoate
COC(=O)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 57.0°C to 64.0°C
Color Cream to White or Green
Boiling Point 265.0°C
Flash Point 144°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HCOC6H4CO2CH3
Quantity 10 g
Solubility Information Solubility in water: insoluble
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Methyl 4-formylbenzoate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'