missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-2-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 15439878
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
174403-69-1
C9H9FO2
168.17
MFCD06203790
ZQFCTCYDRQFPBU-UHFFFAOYSA-N
benzoic acid, 4-fluoro-2-methyl-, methyl ester, 4-fluoro-2-methylbenzoic acid methyl ester, rarechem al bf 0506, methyl=4-fluoro-2-methylbenzoate, 4-fluoro-2-methyl benzoic acid methyl ester, 4-fluoro-2-methyl-benzoic acid methyl ester, benzoic acid,4-fluoro-2-methyl-,methyl ester
2782181
methyl 4-fluoro-2-methylbenzoate
COC(=O)C1=C(C)C=C(F)C=C1
This item is not returnable. View return policy
174403-69-1
C9H9FO2
168.17
MFCD06203790
ZQFCTCYDRQFPBU-UHFFFAOYSA-N
benzoic acid, 4-fluoro-2-methyl-, methyl ester, 4-fluoro-2-methylbenzoic acid methyl ester, rarechem al bf 0506, methyl=4-fluoro-2-methylbenzoate, 4-fluoro-2-methyl benzoic acid methyl ester, 4-fluoro-2-methyl-benzoic acid methyl ester, benzoic acid,4-fluoro-2-methyl-,methyl ester
2782181
methyl 4-fluoro-2-methylbenzoate
COC(=O)C1=C(C)C=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 5 g
Formula Weight 168.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 4-fluoro-2-methylbenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'