missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(chloromethyl)benzoate, 97%, Thermo Scientific™

Catalog Number p-3559326
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
34040-64-7
MFCD00511224
SATDLKYRVXFXRE-UHFFFAOYSA-N
methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester
36664
methyl 4-(chloromethyl)benzoate
COC(=O)C1=CC=C(C=C1)CCl
This item is not returnable. View return policy
34040-64-7
MFCD00511224
SATDLKYRVXFXRE-UHFFFAOYSA-N
methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester
36664
methyl 4-(chloromethyl)benzoate
COC(=O)C1=CC=C(C=C1)CCl
Melting Point 37.0°C to 40.0°C
Color White
Boiling Point 160.0°C to 165.0°C (30.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9ClO2
Linear Formula ClCH2C6H4C(O)OCH3
Quantity 1 g
Molecular Weight (g/mol) 184.62
Formula Weight 184.62
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 4-(chloromethyl)benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-(chloromethyl)benzoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'