missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloro-3-methylbenzoate, 97%, Thermo Scientific™

Catalog Number p-7046093
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
91367-05-4
C9H9ClO2
184.619
MFCD00151838
QOTGNXMPQNGYDM-UHFFFAOYSA-N
4-chloro-3-methylbenzoic acid methyl ester, benzoic acid, 4-chloro-3-methyl-, methyl ester, pubchem3710, acmc-209r8w, 2-chloro-5-methoxycarbonyl toluene, methyl 4-chloranyl-3-methyl-benzoate, 4-chloro-m-toluic acid methyl ester, 4-chloro-3-methylbenzoic acid, methyl ester
7015786
methyl 4-chloro-3-methylbenzoate
CC1=C(C=CC(=C1)C(=O)OC)Cl
This item is not returnable. View return policy
91367-05-4
C9H9ClO2
184.619
MFCD00151838
QOTGNXMPQNGYDM-UHFFFAOYSA-N
4-chloro-3-methylbenzoic acid methyl ester, benzoic acid, 4-chloro-3-methyl-, methyl ester, pubchem3710, acmc-209r8w, 2-chloro-5-methoxycarbonyl toluene, methyl 4-chloranyl-3-methyl-benzoate, 4-chloro-m-toluic acid methyl ester, 4-chloro-3-methylbenzoic acid, methyl ester
7015786
methyl 4-chloro-3-methylbenzoate
CC1=C(C=CC(=C1)C(=O)OC)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.537
Quantity 2 g
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material Methyl 4-chloro-3-methylbenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'