missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals
17.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Melting Point | 54°C to 56°C |
Quantity | 1 g |
Beilstein | 473047 |
Synonym | methyl 4-bromopicolinate, 4-bromo-pyridine-2-carboxylic acid methyl ester, methyl 4-bromo-2-pyridinecarboxylate, methyl4-bromopicolinate, 2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, pubchem12733, acmc-209hag, methyl 4-bromo-pyridine-2-carboxylate |
Solubility Information | Slightly soluble in water. |
Formula Weight | 216.03 |
Percent Purity | ≥97% |
Physical Form | Solid |
Chemical Name or Material | Methyl 4-bromopyridine-2-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'