missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-3-methylbenzoate, 97%, Thermo Scientific Chemicals

Catalog Number 11448810
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
148547-19-7
C9H9BrO2
229.073
MFCD00673014
GTZTYNPAPQKIIR-UHFFFAOYSA-N
4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970
3294148
methyl 4-bromo-3-methylbenzoate
CC1=C(C=CC(=C1)C(=O)OC)Br
This item is not returnable. View return policy
148547-19-7
C9H9BrO2
229.073
MFCD00673014
GTZTYNPAPQKIIR-UHFFFAOYSA-N
4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970
3294148
methyl 4-bromo-3-methylbenzoate
CC1=C(C=CC(=C1)C(=O)OC)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 39°C to 44°C
Quantity 1 g
Beilstein 6593534
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl 4-bromo-3-methylbenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'