Learn More
Thermo Scientific Maybridge Methyl 4-aminothiophene-3-carboxylate, 10g, 95%
Brand: Thermo Scientific Maybridge GK01910EA
Methyl 4-aminothiophene-3-carboxylate, 10g, 95%
Code : 31
Additional Details : CAS Number : 69363-85-5 Weight : 0.06000kg