missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminosalicylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7059784
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
4136-97-4
C8H9NO3
167.164
MFCD00088091
QQOXBFUTRLDXDP-UHFFFAOYSA-N
methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
77787
CHEBI:35089
methyl 4-amino-2-hydroxybenzoate
COC(=O)C1=C(C=C(C=C1)N)O
This item is not returnable. View return policy
4136-97-4
C8H9NO3
167.164
MFCD00088091
QQOXBFUTRLDXDP-UHFFFAOYSA-N
methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
77787
CHEBI:35089
methyl 4-amino-2-hydroxybenzoate
COC(=O)C1=C(C=C(C=C1)N)O

Methyl 4-aminosalicylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 167.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 4-aminosalicylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'