missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[3-(dimethylamino)propoxy]benzoate, 97%, Thermo Scientific™

Catalog Number p-3527265
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
190660-97-0
C13H19NO3
237.30
MFCD06203847
SRXQDDFKGDLAJM-UHFFFAOYSA-N
methyl 4-3-dimethylamino propoxy benzoate, 4-3-dimethylamino-propoxy-benzoic acid methyl ester, methyl 4-3-n,n-dimethylamino-1-propyloxy benzoate, methyl-4-3-n,n-dimethylamino-1-propyloxy benzoate
18508114
methyl 4-[3-(dimethylamino)propoxy]benzoate
COC(=O)C1=CC=C(OCCCN(C)C)C=C1
This item is not returnable. View return policy
190660-97-0
C13H19NO3
237.30
MFCD06203847
SRXQDDFKGDLAJM-UHFFFAOYSA-N
methyl 4-3-dimethylamino propoxy benzoate, 4-3-dimethylamino-propoxy-benzoic acid methyl ester, methyl 4-3-n,n-dimethylamino-1-propyloxy benzoate, methyl-4-3-n,n-dimethylamino-1-propyloxy benzoate
18508114
methyl 4-[3-(dimethylamino)propoxy]benzoate
COC(=O)C1=CC=C(OCCCN(C)C)C=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material Methyl 4-[3-(dimethylamino)propoxy]benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'