missing translation for 'onlineSavingsMsg'
Learn More

α-Methyl-3-(trifluoromethyl)benzyl alcohol, 96%, ACROS Organics™

Brand:  Acros Organics 319730050

Code : 29

Additional Details : CAS Number : 454-91-1 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10225811

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

α-Methyl-3-(trifluoromethyl)benzyl alcohol
454-91-1
100.0
96%
CF3C6H4CH(CH3)OH
MFCD00021868
96%
95.0
≥95%
C9H9F3O
190.17

Specifications

α-Methyl-3-(trifluoromethyl)benzyl alcohol
5g
454-91-1
100.0
Authentic
1.4570 to 1.4590 (20°C, 589nm)
≥95%
C9H9F3O
190.17
Clear colorless to slightly yellow
96%
95.0
1.2400g/mL
Glass bottle
Liquid
96%
CF3C6H4CH(CH3)OH
MFCD00021868
Videos
SDS